Learn More
Epredia™ Specimen Stages for HM 520, 525, 550 and 560
Use Epredia™ Specimen Stages for Epredia™ HM 520, 525, 550 and 560 Series with HM 520, 525, 550 and 560 Cryostats..
Brand: Epredia™ 715710
574.74 NOK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 0.10000kg
Specifications
30mm | |
Specimen Stage | |
HM 550 Series |