Learn More
OHAUS™ Specialty Water Bath Clamp
Shop All Ohaus Products
Click to view available options
Size:
Small
Large
Unit Size:
Each
Description
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
- Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects.
- Nickel-plated zinc clamps offer high-tensile strength at an economical price.
Specifications
Specifications
| Type | Specialty Water Bath Clamp |
| Material | Nickel-plated |
| For Use With (Equipment) | Water Bath |
| Description | Clamp, Specialty Water Bath, CLS-WBATHZS |
| Length (English) | 3.50 in. |
| Length (Metric) | 89.00 mm |
| Size | Small |
| Size (Metric) Grip | 5 to 10 mm |
| Jaw Type | 2-Prong Dual Adjust Water Bath |