Learn More
3M™ Specialty Particulate Respirator 9906
Description
- Provides FFP1 (NPF 4) protection against dusts and mists
- Activated carbon layer helps guard against nuisance-level hydrogen fluoride
- Convex shape, nose clip and twin strapping helps provide a comfortable fit across a broad range of face shapes
- Lightweight, cup-shaped design is sturdy and collapse resistant
- Braided, color-coded headbands help you identify the protection level offered: yellow for FFP1
- Designed to work well with 3M™ Eyewear and Hearing Protection products
- Good fit over a wide range of face sizes provided by convex shape, nose clip and twin-strap design
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 149: 2001 + A1: 2009 FFP1 NR D |
| Color | White |
| Filter Type | FFP1 Protection Level |
| Product Type | Specialty Particulate Respirator |
| With Exhalation Valve | No |
| Resistant To | <TLV for Hydrogen Fluoride, Up to 4 x TLV for Particulates |