Learn More
OHAUS™ Specialty Open Ring
Product Code. p-7158952 Shop All Ohaus Products
Click to view available options
Size:
102 x 102 mm Platform
127 x 127 mm Platform
76 x 76 mm Platform
Unit Size:
Each
Product Code. 15702399
Brand: OHAUS™ 30392346
Only null left
Add to basket
Add to basket
OHAUS™ Specialty Open Ring designed to securely grip and position laboratory glassware.
  • Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
  • Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects.
TRUSTED_SUSTAINABILITY
Specifications
Type Open Ring
Material PVC
Description Clamp, Specialty Open Ring, CLS-OPENRPS
Diameter (Metric) 9 mm
Length (English) 12.91 in.
Length (English) Arm 10.000 in.
Length (Metric) 382.00 mm
Length (Metric) Arm 254 mm
Size 76 x 76 mm Platform