Learn More
Hellma™ Special Optical Glass Micro Absorption Cells
Click to view available options
Capacity (Metric):
20,000 μL
Unit Size:
Each
Specifications
Specifications
| Type | Micro Absorption Cell |
| Capacity (Metric) | 20,000 μL |
| Material | Special Optical Glass |
| Width (Metric) Interior | 2mm |
| Path Length | 10 mm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |