Learn More
SPC25 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
Antigen SPC25
Concentration 0.3 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16844935
View Documents
Proteintech
26474-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
16834935
View Documents
Proteintech
26474-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Description
Description
This gene encodes a protein that may be involved in kinetochore-microtubule interaction and spindle checkpoint activity.
Specifications
Specifications
SPC25
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9HBM1
57405
SPC25 Fusion Protein Ag23985
Primary
-20°C
SPC25
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
AD024, hSpc25, Kinetochore protein Spc25, SPBC25, SPC25
SPC25
IgG
Antigen Affinity Chromatography