Learn More
SPC24 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26268-1-AP-20UL
This item is not returnable.
View return policy
Description
The SPC24 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SPC24. This antibody recognizes human antigen. The SPC24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SPC24 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 147841 | |
| IgG |
| Western Blot | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human |