Learn More
SPC24 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26268-1-AP-20UL
 View more versions of this product
Product Code. 16804785
  • 1710.33 NOK / 20µL
Description
The SPC24 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SPC24. This antibody recognizes human antigen. The SPC24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SPC24
Polyclonal
Unconjugated
20 μL
147841
IgG
Western Blot
267 μg/mL