Learn More
Brady™ SPC Vertical Roll Dispenser
Product Code. 16275492 Shop All Brady Products
Change view
Click to view available options
For Use With (Equipment):
1 meter roll
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16275492 1 meter roll 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16275492 Supplier Brady™ Supplier No. 830954
  • ONLINE EXCLUSIVE PRICE
    3784.00 NOK
    3093.05 NOK / Pack of 1
    Save 690.95 NOK
    18% Off
Estimated Shipment: 11-01-2027
Features wheels for easy maneuverability
Specifications
Color Yellow
Material Steel
Type SPC Accessories
For Use With (Equipment) 1 meter roll
Packaging Quantity 1 Ea.
Width (Metric) 1 m
Weight (Metric) 7700 g
Alternate Part Number DJR128