Learn More
Brady™ SPC™ Absorbents SitePoint Spill Ready Kit
Product Code. 17821775
Click to view available options
Capacity (English):
26.4 gal.
Unit Size:
1 set
Product Code. 17821775
Brand: Brady™ 309077
Ideal for moving spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required, which is especially ideal for maintenance activities, at construction sites and other temporary locations.
  • Loaded SitePoint Mobile Unit with suitable products for cleaning up general spills
  • Features two (200 mm) wheels for mobility and anchor points for fixed installation
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
  • Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission
Specifications
Color Yellow, White
Capacity (English) 26.4 gal.
Capacity (Metric) 100 L
Material MDPE
Type Spill Kit
Format Spill Kit
For Use With (Application) Beverage and Food Processing, Chemical, Healthcare, Maritime, Oil and Gas, Pharmaceutical, Warehouse
Includes SitePoint Mobile Unit, SPK Scoop, Economy Maintenance Spill Kit, Spillfix, Floor Stand, Fixings
Length (English) 2.3 in.
Length (Metric) 5.95 cm
Width (English) 29.724 in.
Width (Metric) 75.5 cm
Height (Metric) 172.5 cm
Depth (Metric) 5.95 cm
Weight (English) 35.3 lb.
Weight (Metric) 16 kg
Alternate Part Number SPHS1-BB1-YE-SPILL