Learn More
Brady™ SPC™ Absorbents Battery Acid Specialty Spill Kit
Description
Position the battery acid spill kits near a battery swap station and around your facility so that you are prepared should a spill occur.
- This spill kit is designed to pick up battery acid on both smooth and rough surfaces while remaining chemically inert.
- These kits combine pads, spillfix, PPE and accessories so that you are prepared in an emergency.
- This Spill kit contains chemical neutralizing polymers which provide safe spill control with a color change indicator that reflects neurtalisation.
Specifications
Specifications
| Capacity (English) | 5.283 gal. |
| Type | Spill Kit |
| Includes | 2 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper, 1 x SF-JAR-CS (149265) |
| Model | Acids |
| Alternate Part Number | SKH-BATT-E |