Learn More
Brady™ SPC™ Absorbents ALLWICK Universal FLEXISOC
Description
AW561 is an universal sorbent soc, filled with universal meltblown polypropylene that absorbs industrial liquids including oils, water, solvents, coolants, etc.
- The ultimate soc for flexibility: cut the soc into the length you want and re-clip. No need to stock multiple lengths.
- Strong wicking action: absorption begins instantly after contact with the spilled liquid, no need to turn the soc.
- Flexible design allows the soc to fit around any machine, corner or curve.
- The ideal sorbent for containing and absorbing leaks and spills around machines.
Specifications
Specifications
| Color | Gray |
| Capacity (English) | 13.737 gal. |
| Capacity (Metric) | 52 L |
| Material | Polypropylene |
| Type | Absorbent Roll |
| For Use With (Application) | Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Warehouse and Logistics |
| Model | Universal Sorbent |
| Length (Metric) | 17 m |
| Diameter (English) | 2.992 in. |
| Diameter (Metric) | 7.60 cm |
| Show More |