missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26142-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| SPATS1 | |
| Polyclonal | |
| Unconjugated | |
| SPATS1 | |
| SPATA8, SPATS1, SRSP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221409, 71020 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2RRY8, Q496A3 | |
| Spats1 | |
| SPATS1 Fusion Protein Ag23533 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction