Learn More
SI Analytics™ Spare Tube for TitroLine™ KF and TitroLine™ KF trace
Product Code. 10314971 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric):
1 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10314971 1 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10314971 Supplier SI Analytics™ Supplier No. 285226115
  • ONLINE EXCLUSIVE PRICE
    216.00 NOK
    201.70 NOK / Pack of 1
    Save 14.30 NOK
    6% Off
In Stock
Spare / accessory item
  • 1 m length tube
TRUSTED_SUSTAINABILITY
Specifications
Description Spare tube for TitroLine KF and TitroLine KF trace
Diameter (Metric) 3 x 0.5 mm
Length (Metric) 1 m