Learn More
IKA™ Spare Stainless Steel Cutter
Shop All IKA Products
Click to view available options
For Use With (Application):
For Crushing Materials up to Mohs Hardness 5
Unit Size:
Each
Specifications
Specifications
| Product Type | Spare Cutter |
| For Use With (Application) | For Crushing Materials up to Mohs Hardness 5 |
| Hardness | Up to 5 °Mols |
| Material | Stainless Steel |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering