Learn More
Buerkle™ Spare Spray Heads for Buerkle™ Spray Bottle
Product Code. p-7230562 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11818000 250 mL Each
13471379 500 mL Each
10452080 1000 mL Each
3 options
Product Code. 11818000
Brand: Buerkle™ 03092025
Only null left
Add to basket
Add to basket
Manufactured from polypropylene and polyethylene. Buerkle™ Spray Heads for Spray Bottle has a lever pump sprayer with adjustable nozzle.
  • Manufactured from polypropylene and polyethylene
  • Lever pump with adjustable nozzle
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Volume (Metric) Dispense 1.2 mL/Stroke
Closure Color White
Product Type Lever Pump Sprayer with Adjustable Nozzle
For Use With (Equipment) Spray Bottle
Closure Material LDPE, Polypropylene, Stainless Steel
Closure Type Spray
Through the Closure Dispensing Yes