Learn More
BRAND™ Spare Key for Lateral Burette Stopcock
Product Code. 10380092 Shop All Brand Products
Change view
Click to view available options
:
For Use With (Equipment): 25 mL Burette
Product Code.
10380092 For Use With (Equipment): 25 mL Burette
1 options
Product Code. 10380092
Brand: BRAND™ 81141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stopcock housing with burette tip. Interchangeable borosilicate glass key with retention device.
  • Securing Device
  • Boro 3.3
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity (Metric) 25 mL
    For Use With (Equipment) 25 mL Burette
    Material Borosilicate Glass
    Includes Retention Device
    Type Stopcock Key