Learn More
SPARC Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Product Code. 16894374 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16894374 20 μL 20µL
16884374 150 μL 150µL
2 options
Product Code. 16894374
Brand: Proteintech 152741AP20UL
Rabbit Polyclonal Antibody
SPARC is a secreted protein, acidic and rich in cysteines. It is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM)
Specifications
Antigen SPARC
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPARC
Gene Accession No. P07214, P09486, P16975, P20112
Gene Alias Basement membrane protein 40, BM 40, ON, Osteonectin, SPARC
Gene Symbols SPARC
Host Species Rabbit
Immunogen SPARC Fusion Protein Ag7390
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20692, 24791, 595124, 6678
Target Species Human, Mouse, Rat, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG