missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Span™ 20, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is 100% bio-based non-ionic surfactant, thus aligns with Use of Renewable Feedstocks and belongs to 12 principles aligned category of our greener alternatives.
Specifications
Specifications
| Boiling Point | >100°C/1013 hPa |
| Density | 1.0 g/cm3 at 20°C |
| Concentration | 100% |
| Quantity | 500 mL |
| MDL Number | MFCD00005365 |
| Synonym | Span™ 20, Sorbitan monolaurate, Sorbitan monododecanoate, Span™ 20 |
| Molecular Weight (g/mol) | 346.46 |
| Grade | Ungraded |
| Chemical Name or Material | Span™ 20 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?