missing translation for 'onlineSavingsMsg'
Learn More

SPAG8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13915-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892334

  • 1281.92 NOK / 20µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein encoded by this gene is recognized by sperm agglutinating antibodies from an infertile woman. This protein is localized in germ cells of the testis at all stages of spermatogenesis and is localized to the acrosomal region of mature spermatozoa. Alternatively spliced variants that encode different protein isoforms have been described but the full-length sequences of only two have been determined.
Specifications

Specifications

SPAG8
Polyclonal
Unconjugated
SPAG8
BS 84, HSD 1, hSMP 1, SMP 1, SMP1, SPAG3, SPAG8, sperm associated antigen 8, Sperm membrane protein 1, Sperm membrane protein BS 84
Rabbit
Antigen Affinity Chromatography
RUO
26206, 433700
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3V0Q6, Q99932
Spag8
SPAG8 Fusion Protein Ag4910
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers