missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SP5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16669954 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
16669954 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 16669954

Brand: Invitrogen™ PA5103505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SP5.

SP5 binds to GC boxes promoters elements. SP5 probable transcriptional activator that has a role in the coordination of changes in transcription required to generate pattern in the developing embryo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SP5
Gene Accession No. Q6BEB4, Q9JHX2
Gene Alias SP5; Sp5 transcription factor; trans-acting transcription factor 5; transcription factor Sp5
Gene Symbols SP5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SP5(Accession Q6BEB4), corresponding to amino acid residues T260-T310.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389058, 64406
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.