Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
1.25 mL
2.5 mL
5 mL
8 mL
Unit Size:
Pack of 10
Description
- Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
- Comes with flat bottom shape to allow use as weighing boat
- Available in white color
- Available in individual packs for one time use
- Includes certificate of processing for sterilization
Specifications
Specifications
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Shape | Flat Bottom |
| Capacity (Metric) | 8 mL |
| Color | White |
| Certifications/Compliance | FDA |
| Disposable | Single-use |
| Product Type | Spoon |
| For Use With (Application) | Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders |
| Includes | Certificate of processing for sterilization |
| Show More |