Learn More
Bel Art™ Poxygrid™ Rack and a Half
Product Code. 15560042 Shop All SP Scienceware Products
Click to view available options
Positions:
100
180
Array:
5 x 20
9 x 20
Dimensions (L x W x H):
38.6 x 10.8 x 6.4 cm
38.6 x 18.3 x 6.4 cm
Unit Size:
Each
Product Code. 15560042
Brand: Bel Art™ F187860770
Only null left
Add to basket
Add to basket
Extra-long wire test tube racks for high volume test tube work
  • High quality epoxy-coated wire racks
  • Epoxy coated steel wire is chemical and corrosion resistant
  • Steam autoclavable at 121°C (250°F)
Specifications
Product Type Test Tube Rack
Array 9 x 20
Autoclavable Autoclavable
Color Blue
Holds Tubes (13 to 16 mm)
Material Steel Wire
Positions 180
Shape Rectangular
Shelves 3 Tiers
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) 13 - 16mm Tubes
Temperature (Metric) Range -223°C to +121°C
Dimensions (L x W x H) 38.6 x 18.3 x 6.4 cm
Length (Metric) 38.6 cm
Length (English) 15.18 in.
Width (Metric) 18.3 cm
Width (English) 7.19 in.
Height (Metric) 6.4 cm
Height (English) 2.5 in.
Diameter (Metric) Opening 16 mm