Learn More
Bel-Art™ SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
Product Code. p-8899014 Shop All SP Scienceware Products
Click to view available options
Product Type:
Replacement Blade
Unit Size:
Each
Product Code. 10476834
Brand: Bel-Art™ H210110000
Only null left
Add to basket
Add to basket
Portable, pocket-sized cutter cuts flexible tubing from 1.6mm to 19mm (1/16 to 3/4 in.) O.D.
  • Hardened steel replacement blade for use with the Bel-Art™ Pocket-Sized Tubing Cutter 22-088-245
  • Makes clean, straight cuts
Specifications
Material Steel
Overall Length (English) 3 in.
Overall Length (Metric) 76.2 mm
For Use With Tubing Cutter
Product Type Replacement Blade
Not recommended for latex tubing.