Learn More
3M™ Sound Examiner 400 Series Sound Level Meter SE 401-IS
Product Code. 15730038
Change view
Click to view available options
Battery Life:
18 hours
Unit Size:
Each
Product Code. Battery Life unitSize
15730038 18 hours Each
1 options
Product Code. 15730038
Brand: 3M™ 7100015831
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Sound Examiner 400 Series Sound Level Meter SE 401-IS is an intrinsically safe meter engineered to accurately measure noise levels in highly variable environments.
  • Intrinsically safe
  • SPL, LMAX, LMIN, Peak, LEQ/LAVG, Elapsed Time
  • Variable, user-selectable data logging intervals
  • Rechargeable lithium battery
  • Store numerous noise surveys in memory for download
  • Removable Type/Class 1 microphone
  • Class 1
  • Multilingual user interface
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 18 hours
Dimensions 28.4 x 7.1 x 3.3 cm
Product Type Sound Level Meter
For Use With (Application) Area Monitoring (Sound Level Meter)