missing translation for 'onlineSavingsMsg'
Learn More

Sotuletinib, MedChemExpress

Product Code. p-200098296
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16488047 5 mg 5mg
16425976 100 mg 100mg
16421537 50 mg 50mg
16415338 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16488047

Brand: Medchem Express HY12768

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs.

Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs.

Specifications

Chemical Name or Material Sotuletinib
CAS 953769-46-5
Color Light Brown
Purity Grade Notes Research
Molecular Formula C20H22N4O3S
Quantity 5 mg
Synonym BLZ945
Solubility Information DMSO : 83.33 mg/mL (209.12 mM; Need ultrasonic)
SMILES O=C(C1=NC=CC(OC2=CC=C3N=C(N[C@H]4[C@H](O)CCCC4)SC3=C2)=C1)NC
Molecular Weight (g/mol) 398.48
Formula Weight 398.48
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.