Learn More
Sotuletinib, MedChemExpress
Product Code. 16488047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16488047
Brand: Medchem Express HY12768
MedChemExpress Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs.
Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,000-fold selectivity against its closest receptor tyrosine kinase homologs.
Specifications
Chemical Name or Material Sotuletinib
CAS 953769-46-5
Color Light Brown
Purity Grade Notes Research
Molecular Formula C20H22N4O3S
Quantity 5 mg
Synonym BLZ945
Solubility Information DMSO : 83.33 mg/mL (209.12 mM; Need ultrasonic)
SMILES O=C(C1=NC=CC(OC2=CC=C3N=C(N[C@H]4[C@H](O)CCCC4)SC3=C2)=C1)NC
Molecular Weight (g/mol) 398.48
Formula Weight 398.48
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.