Learn More
Thermo Scientific™ Sorvall™ Microtube Adapters
Product Code. p-4527834 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
0.2mL microtubes
0.5/0.6mL microtubes
For 0.25/0.4mL microtubes
Unit Size:
Pack of 24
Product Code. For Use With (Equipment) unitSize
12883312 0.2mL microtubes Pack of 24
11828362 0.5/0.6mL microtubes Pack of 24
11818362 For 0.25/0.4mL microtubes Pack of 24
3 options
Product Code. 12883312
Brand: Thermo Scientific™ 76003750
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ adapters.
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.
Ordering Information:
Sold in packs of 24
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 0.2mL microtubes
Color Gray
No. per Pack 24