Learn More
Thermo Scientific™ Sorvall™ Microtube Adapters
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
0.2mL microtubes
0.5/0.6mL microtubes
For 0.25/0.4mL microtubes
Unit Size:
Pack of 24
Description
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.
Ordering Information:
Sold in packs of 24
Specifications
Specifications
| For Use With (Equipment) | 0.5/0.6mL microtubes |
| Color | Turquoise |
| No. per Pack | 24 |