Description
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.
Ordering Information:
Sold in packs of 24
Specifications
Specifications
| For Use With (Equipment) | For 0.25/0.4mL microtubes |
| Description | For 0.25/0.4mL microtubes; Red |
| Color | Red |