Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Shop All Thermo Scientific Products
Click to view available options
Capacity:
1 x 250 mL
1 x 750 mL
Unit Size:
Each
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
Specifications
Specifications
| For Use With (Equipment) | Swinging bucket rotor |
| Capacity | 1 x 250 mL |
| Type | Bucket |
| Description | Rectangular Bucket |