Learn More
Sonoco™ ThermoSafe PolarPack™ MoistureGuard™
Description
- Outer film layer assures products arrive in top condition by absorbing condensation.
- Provide refrigeration for transport of food, pharmaceuticals, and medical products.
- Condition coolant in freezer before shipments.
- Available in several sizes
Specifications
Specifications
| For Use With (Application) | For shipping temperature critical packages during transit |
| Freezing Point | 0°C |
| Material | Polyethylene |
| Weight (English) | 16 oz. |
| Weight (Metric) | 0.45 kg |
| Dimensions (L x W x H) | 6.5 x 5.5 x 0.75 in. |