Learn More
Branson Ultrasonics™ Sonifier™ Disruptor Horns
Description
For use with Sonifier S-250 and S-450 Cell Disruptors
Transmit ultrasonic energy into a solution.
Titanium construction
High Gain Horn
Step Horn with Threaded Body (22-020-860)
- Tapped horn end enables attachment of microtips
- Includes removable flat tip
- Can be used with screw-on accessories, such as a continuous flow cell
Provides higher amplitude at 0.75 and 1 in. diameters (1.9 and 2.5cm) than similar step horns
Specifications
Specifications
| Amplitude | 21 to 145 μm |
| Product Type | Disruptor Horn |
| Volume Range (Metric) | 10 to 250mL |
| Diameter (English) Tip | 1/2 in. |
| Diameter (Metric) Tip | 13 mm |
| Material | Titanium |
| For Use With (Application) | Ideal for transmit ultrasonic energy into a solution |
| For Use With (Equipment) | Sonifier™ S-250 and S-450 cell disruptor |
| Intensity | High |