Learn More
Sonidegib, MedChemExpress
Product Code. p-200099337
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16483127
Brand: Medchem Express HY16582A
MedChemExpress Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.
Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.
Specifications
Chemical Name or Material Sonidegib
CAS 956697-53-3
Color Purple
Purity Grade Notes Research
Molecular Formula C26H26F3N3O3
Quantity 50 mg
Synonym Erismodegib LDE225 NVP-LDE225
Solubility Information DMSO : 50 mg/mL (102.99 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C(C(C=C2)=CC=C2OC(F)(F)F)=CC=C1)NC3=CC=C(N=C3)N4C[C@@H](C)O[C@@H](C)C4
Molecular Weight (g/mol) 485.5
Formula Weight 485.5
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.