Learn More
Cytiva Sonicated DNAs for Hybridization Studies
Product Code. 10733217 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Sonicated Calf Thymus DNA
Sonicated Salmon Sperm DNA
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10733217 Sonicated Salmon Sperm DNA Each
10723217 Sonicated Calf Thymus DNA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10733217 Supplier Cytiva Supplier No. 27456501
  • ONLINE EXCLUSIVE PRICE
    5310.00 NOK
    5017.00 NOK / Pack of 1
    Save 293.00 NOK
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.
  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp
TRUSTED_SUSTAINABILITY
Specifications
Format Solid
Product Type Sonicated Salmon Sperm DNA
Content And Storage -20°C
Quantity 100/Pk.
For Use With (Application) Hybridisation experiments