Learn More
Sonic Hedgehog/Shh N-Terminus Rat anti-Mouse, Clone: 171018, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB4641-SP
Encompass
Product Code. 15747034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
Sonic Hedgehog/Shh
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
6469
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Western Blot, Immunohistochemistry, Neutralization, ELISA
171018
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q62226
SHH
E. coli-derived recombinant mouse Sonic Hedgehog Cys25-Gly198 (Lys122Arg) Accession # Q62226
25 μg
Primary
Detects mouse Sonic Hedgehog in ELISAs. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Dhh (N-terminus; aa 23-198) is observed and no cross-reactivity with rmIhh (N-terminus; aa 66-240), rmDhh (C-terminus; aa 199-396), or rmShh (C-terminus; aa 199-437) is observed.
Mouse