missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solvent blue 35, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
Specifications
Specifications
| Formula Weight | 350.5 |
| Density | 1.23g/mL |
| Quantity | 50 g |
| Physical Form | Powder |
| Chemical Name or Material | Solvent Blue 35, Dye content: Approx. 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction