Learn More
3M™ Solus™ Safety Glasses
Description
- Lens curve has been calculated to optimize the fitting, field of vision and coverage of the eyes
- Extremely stylish and lightweight glasses
- Soft inner padding of the temple, leading to increased comfort
- Supplied with a microfiber storage bag for cleaning and protection
- Anti-scratch and anti-fog coating for increased durability and improved vision
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 166: 2001 |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety glasses |
| Frame Color | Black, Orange |
| Includes | Microfiber storage bag |
| Lens Material | Polycarbonate |