Learn More
Sonics Materials™ Solid Probe
Click to view available options
Diameter (Metric):
13 mm
19 mm
25 mm
Unit Size:
Each
Specifications
Specifications
| Amplitude | 35 μm |
| Product Type | Dispersing Aggregate |
| Diameter (Metric) | 25 mm |
| Length (Metric) | 122 mm |
| Intensity | Low |
| Packaging Quantity | 1 Ea. |