Learn More
Sonics Materials™ Solid Probe
Specifications
Specifications
| Amplitude | 58 μm |
| Product Type | Digital Disperser |
| Diameter (Metric) | 19 mm |
| Length (Metric) | 127 mm |
| Intensity | Medium |
| Packaging Quantity | 1 Ea. |