Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Clamp Spare Kit (Screws)
Dual Stack Universal Platform
Dual Stack Universal Platform Upgrade Kit
Platform Spare Part Kit
Screwdriver
Set of O-rings
Universal Platform
For Use With (Equipment):
Solaris 2000
Solaris 2000 and 4000
Solaris 4000
Solaris Open Air Shakers and Temperature Controlled Shakers, MaxQ 6000 Shakers
Dimensions (L x W):
6 x 6 in.
260 x 25 mm
12 x 14 in.
18 x 18 in.
18 x 24 in.
18 x 30 in.
36 x 24 in.
250 x 150 mm
Unit Size:
1 unit
Each
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab. Select a platform and clamp configuration that best meets your application, whether you require a single vessel type and size.
Certifications
UL, cUL, CE
Warranty
1 Year
Specifications
Specifications
| Product Type | Screwdriver |
| Dimensions (L x W) | 260 x 25 mm |
| For Use With (Equipment) | Solaris Open Air Shakers and Temperature Controlled Shakers, MaxQ 6000 Shakers |