Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Shop All Thermo Scientific ProductsDescription
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Warranty
1 Year
Specifications
Specifications
| Product Type | Dual Stack Universal Platform |
| Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| Dimensions (L x W) | 18 x 18 in. |
| Length (English) Platform | 18 in. |
| Length (Metric) Platform | 457.2 mm |
| Width (English) Platform | 18 in. |
| Width (Metric) Platform | 457.2 mm |
| For Use With (Equipment) | Solaris 2000 |
| Includes | Two platforms, screws, and supporting rods |
| Product Line | Solaris |
| Show More |