Learn More
Thermo Scientific™ SOLA™ SPE Plates
Product Code. 15550163 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
10 mg/2 mL
30 mg/2 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Mixed Mode Weak Anion Exchanger, Polymeric
Mixed Mode Weak Cation Exchanger, Polymeric
Unit Size:
1 pack
Each
Product Code. Description Sorbent unitSize
15550163 10 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric Each
11899163 10 mg/2 mL HRP (Reversed Phase Polymeric) Each
16697845 30 mg/2 mL HRP (Reversed Phase Polymeric) 1 pack
11809173 10 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric Each
15547495 10 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric Each
16607855 30 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric 1 pack
11819173 10 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric Each
16617855 30 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric 1 pack
13493038 10 mg/2 mL Mixed Mode Weak Cation Exchanger, Polymeric Each
16627855 30 mg/2 mL Mixed Mode Weak Cation Exchanger, Polymeric 1 pack
15179653 10 mg/2 mL Mixed Mode Weak Anion Exchanger, Polymeric Each
16637855 30 mg/2 mL Mixed Mode Weak Anion Exchanger, Polymeric 1 pack
12 options
Product Code. 15550163
Brand: Thermo Scientific™ 60309003
Only null left
Add to basket
Add to basket
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
SOLA Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You’ll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
10mg/2mL 96 well plate format
Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
SOLA products are also available in cartridge format.
TRUSTED_SUSTAINABILITY
Specifications
Description 10 mg/2 mL
Bed Weight 10 mg
For Use With (Application) Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Acidic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Product Line SOLA
Sorbent Mixed Mode Strong Anion Exchanger, Polymeric
Type 96-well SPE Plate
Volume (Metric) 2 mL
Volume (Metric) Well 2 mL
Quantity 1 Pk.