Description
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
- Higher levels of reproducibility
- Lower elution volumes
- High extract cleanliness
10mg/2mL 96 well plate format
Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
SOLA products are also available in cartridge format.
Specifications
Specifications
| Description | 10 mg/2 mL |
| Bed Weight | 10 mg |
| For Use With (Application) | Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Acidic Compounds. Complementary Reversed Phase Retention of Neutral Compounds. |
| Product Line | SOLA |
| Sorbent | Mixed Mode Strong Anion Exchanger, Polymeric |
| Type | 96-well SPE Plate |
| Volume (Metric) | 2 mL |
| Volume (Metric) Well | 2 mL |
| Quantity | 1 Pk. |