Learn More
Diversey™ Soft Care Des E H5 Hand Disinfectant Gel
Product Code. 17986710
Change view
Click to view available options
Volume (Metric):
0.5 L
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17986710 0.5 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17986710 Supplier Diversey™ Supplier No. 101105181
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Soft Care Des E is an alcohol based (71.5%) hand disinfectant gel for waterless skin disinfection suitable for use in all areas, including healthcare and food processing environments.
  • Hand disinfectant based on ethanol, a disinfectant which is very effective against resident and transient flora such as bacteria (e.g. MRSA) and yeast
  • Also effective against enveloped viruses, such as influenza and certain non-enveloped viruses such as Norovirus and Adenovirus
  • Contains a skin softener helping the skin to remain smooth and intact, particularly in areas where frequent hand disinfection is required
  • Perfume-free, eliminating the risk of tainting foods and making it suitable for application in food processing areas
  • Skin compatibility dermatologically tested (Patch and TEWL tests)
Specifications
Product Type Disinfectant Gel
Color Clear
Form Gel
Disposable Single-use
Latex Free Yes
Quantity 10 Bottles
Sterility Non-sterile
Volume (Metric) 0.5 L
compliance-icons
Product Identifier
  • Soft Care Des E H5
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, propan-2-ol