missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Gray

Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.

Brand:  Notrax 411C0324GY

Additional Details : Weight : 1.40000kg

 View more versions of this product

Product Code. 17712078

  • 361.00 NOK / Each

Estimated Shipment: 25-10-2023
to see stock.
Add to basket

Description

Description

  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

60 ft.
Industrial and Commercial Matting
60 cm
Gray
Microcellular Vinyl
1830 cm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers