Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Gray
Product Code. p-7227245
Change view
Click to view available options
Length (Metric):
91 cm
150 cm
1830 cm
Width (Metric):
60 cm
91 cm
122 cm
Unit Size:
Each
Product Code. Length (Metric) Width (Metric) unitSize
17732088 91 cm 60 cm Each
17772088 150 cm 91 cm Each
17712078 1830 cm 60 cm Each
17752078 1830 cm 91 cm Each
17792078 1830 cm 122 cm Each
5 options
Product Code. 17732088
Brand: Notrax 411S0323GY
Only null left
Add to basket
Add to basket
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Color Gray
Length (English) 36 in.
Length (Metric) 91 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 60 cm