missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly bottle
Unit Size:
2.5kg
25g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 2.5 kg |
| Linear Formula | Na2S2O3 · 5H2O |
| UN Number | NONH for all modes of transport |
| Solubility Information | Soluble in water |
| Formula Weight | 248.18g/mol |
| Percent Purity | ≥99.5% |
| Grade | ACS Reagent |
| Packaging | Poly bottle |
| Chemical Name or Material | Sodium Thiosulfate Pentahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction