missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, TRC

Product Code. 30010263
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010263 50 g 50g
30132425 10 g 10g
30136706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010263 Supplier Toronto Research Chemicals Supplier No. TRCS67309050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4 O7 . 2 Na . 10 H2 O
Molecular Weight (g/mol) 381.37
Synonym Borax (B4Na2O7.10H2O), Borax (8CI), 20 Mule Team Borax, 20 Mule Team Borax Tech 4/200 Mesh, BR 100, BR 100 (borate), Borascu, Borax decahydrate, Boric acid (H2B4O7), disodium salt, decahydrate, Boricin, Boron sodium oxide (B4Na2O7), decahydrate, Bura, Disodium tetraborate decahydrate, Gerstley borate, M 9358, Sodium biborate decahydrate, Sodium borate (Na2B4O7), decahydrate, Sodium pyroborate, Sodium pyroborate decahydrate, Sodium tetraborate decahydrate, Solubor, Solubor DF, Three Elephant, Tincal
IUPAC Name disodium;3,7-dioxido-2,4,6,8,9-pentaoxa-1,3,5,7-tetraborabicyclo[3.3.1]nonane;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1OB2OB([O-])OB(O1)O2

Specifications

Quantity 50 g
InChI Formula InChI=1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;
Formula Weight 382.0868
Chemical Name or Material Borax
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.