Learn More
Sodium Tetraborate Decahydrate, TRC
Product Code. 30132425
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132425 10 g 10g
30136706 1 g 1g
30010263 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132425 Supplier Toronto Research Chemicals Supplier No. TRCS67309010G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1303-96-4
Molecular Formula B4 O7 . 2 Na . 10 H2 O
Molecular Weight (g/mol) 381.37
Synonym Borax (B4Na2O7.10H2O), Borax (8CI), 20 Mule Team Borax, 20 Mule Team Borax Tech 4/200 Mesh, BR 100, BR 100 (borate), Borascu, Borax decahydrate, Boric acid (H2B4O7), disodium salt, decahydrate, Boricin, Boron sodium oxide (B4Na2O7), decahydrate, Bura, Disodium tetraborate decahydrate, Gerstley borate, M 9358, Sodium biborate decahydrate, Sodium borate (Na2B4O7), decahydrate, Sodium pyroborate, Sodium pyroborate decahydrate, Sodium tetraborate decahydrate, Solubor, Solubor DF, Three Elephant, Tincal
IUPAC Name disodium;3,7-dioxido-2,4,6,8,9-pentaoxa-1,3,5,7-tetraborabicyclo[3.3.1]nonane;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1OB2OB([O-])OB(O1)O2
Specifications
Quantity 10 g
InChI Formula InChI=1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;
Formula Weight 382.0868
Chemical Name or Material Borax