Learn More
Sodium tert-Butylate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 865-48-5 |
| Melting Point | 263°C |
| Molecular Formula | (CH3)3CONa |
| MDL Number | MFCD00040575 |
| Quantity | 100 g |
| Synonym | Sodium tert-butylate, Sodium-t-butoxide, Sodium tert-butylate |
| Molecular Weight (g/mol) | 96.1 |
| Grade | Research |